missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ubiquitin-like-conjugating enzyme ATG10 Protein
A DNA sequence encoding the Homo sapiens (Human) Ubiquitin-like-conjugating enzyme ATG10, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP8050-ec-50ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
NP_001124500.1 | |
Ubiquitin-like-conjugating enzyme ATG10 Protein | |
Research Use Only | |
ATG10 | |
Recombinant Protein | |
E. coli | |
MEEDEFIGEK TFQRYCAEFI KHSQQIGDSW EWRPSKDCSD GYMCKIHFQI KNGSVMSHLG ASTHGQTCLP MEEAFELPLD DCEVIETAAA SEVIKYEYHV LYSCSYQVPV LYFRASFLDG RPLTLKDIWE GVHECYKMRL LQGPWDTITQ QEHPILGQPF FVLHPCKTNE FMTPVLKNSQ KINKNVNYIT SWLSIVGPVV GLNLPLSYAK ATSQDERNVP | |
Greater than 80% as determined by SDS-PAGE. |
83734 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |