missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human UCK2 Protein
A cDNA sequence encoding the UCK2 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13880-25ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
UCK2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MAGDSEQTLQ NHQQPNGGEP FLIGVSGGTA SGKSSVCAKI VQLLGQNEVD YRQKQVVILS QDSFYRVLTS EQKAKALKGQ FNFDHPDAFD NELILKTLKE ITEGKTVQIP VYDFVSHSRK EETVTVYPAD VVLFEGILAF YSQEVRDLFQ MKLFVDTDAD TRLSRRVLRD ISERGRDLEQ ILSQYITFVK PAFEEFCLPT KKYADVIIPR GADNLVAINL IVQHIQDILN GGPSKRQTNG CLNGYTPSRK RQASESSSRP HLEHHHHHH | |
Greater than 90% as determined by SDS-PAGE. |
25 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The UCK2 protein solution (1 mg/1ml) is formulated in 20mM Tris-HCl buffer (pH 8.0), 2mM DTT, 100mM NaCl, 0.1mM PMSF, 1mM EDTA and 20% glycerol . |