missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human YAE1D1 Protein
A cDNA sequence encoding the YAE1D1 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13980-1mg
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
YAE1D1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSWVQAA SLIQGPGDKG DVFDEEADES LLAQREWQSN MQRRVKEGYR DGIDAGKAVT LQQGFNQGYK KGAEVILNYG RLRGTLSALL SWCHLHNNNS TLINKINNLL DAVGQCEEYV LKHLKSITPP SHVVDLLDSI EDMDLCHVVP AEKKIDEAKD ERLCENNAEF NKNCSKSHSG IDCSYVECCR TQEHAHSENP SPTWILEQTA SLVKQLGLSV DVLQHLKQL | |
Greater than 95.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The YAE1D1 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 20% glycerol and 1mM DTT. |