missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Zinc finger protein 384 Protein
A DNA sequence encoding the Homo sapiens (Human) Zinc finger protein 384, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP8445-ec-10ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
Zinc finger protein 384 Protein | |
Research Use Only | |
ZNF384 | |
Recombinant Protein | |
E. coli | |
MEESHFNSNP YFWPSIPTVS GQIENTMFIN KMKDQLLPEK GCGLAPPHYP TLLTVPASVS LPSGISMDTE SKSDQLTPHS QASVTQNITV VPVPSTGLMT AGVSCSQRWR REGSQSRGPG LVITSPSGSL VTTASSAQTF PISAPMIVSA LPPGSQALQV VPDLSKKVAS TLTEEGGGGG GGGGSVAPKP PRGRKKKRML ESGLPEMNDP YVLSPEDDDD HQKDGKTYRC RMCSLTFYSK SEMQI | |
Greater than 90% as determined by SDS-PAGE. |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |