missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza-B Qingdao Protein
A cDNA sequence encoding the Influenza-B Qingdao was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12442-50ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
Influenza-B Qingdao Protein | |
Research Use Only | |
Recombinant Protein | |
Influenza B virus | |
The B/Qingdao/102/91 solution contains 0.1M NaCl, 10mM Tris HCl, 1mM EDTA pH 8.0, 0.1% sodium azide (NaN3) and 0.005% thimerosal. |
50 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Influenza | |
Untagged | |
Greater than 90.0% as determined byAnalysis by SDS-PAGE. |