missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse BCA 1 Protein
A cDNA sequence encoding the BCA 1 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP10530-5ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
55985 | |
5 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The ED50, as measured by its ability to chemoattract human CXCR5-transfected mouse BaF3 cells, is less than 2microg/ml. | |
Mouse | |
Untagged | |
The BCA1 protein was lyophilized from a 0.2microm filtered solution in Acetonitrile and TFA. |
BCA 1 Protein | |
Research Use Only | |
Cxcl13 | |
Recombinant Protein | |
E. coli | |
ILEAHYTNLK CRCSGVISTV VGLNIIDRIQ VTPPGNGCPK TEVVIWTKMK KVICVNPRAK WLQRLLRHVQ SKSLSSTPQA PVSKRRAA | |
Greater than 97.0% as determined by SDS-PAGE. |