missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CD55 Protein
A cDNA sequence encoding the CD55 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP11339-2ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
13136 | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
DCGPPPDIPN ARPILGRHSK FAEQSKVAYS CNNGFKQVPD KSNIVVCLEN GQWSSHETFC EKSCVAPERL SFASLKKEYL NMNFFPVGTI VEYECRPGFR KQPPLPGKAT CLEDLVWSPV AQFCKKKSCP NPKDLDNGHI NIPTGILFGS EINFSCNPGY RLVGVSSTFC SVTGNTVDWD DEFPVCTEIH CPEPPKINNG IMRGESDSYT YSQVVTYSCD KGFILVGNAS IYCTVSKSDV GQWSSPPPRC IEKSKVPTKK PTINVPSTGT PSTPQKPTTE SVPNPGDQPT PQKPSTVKVS ATQHVPVTKT TVRHPIRTST DKGEPNTGLE HHHHHH | |
Greater than 95.0% as determined by SDS-PAGE. |
CD55 Protein | |
Research Use Only | |
Cd55 | |
Mouse | |
His | |
CD55 protein solution (0.5 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |