missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse En Protein
A cDNA sequence encoding the En was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP10587-100ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
13805 | |
100 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Measured by its ability to bind with rhTGF-beta RII/Fc in a functional ELISA. Optimal dilutions should be determined by each laboratory for each application. | |
Mouse | |
His | |
Endoglin was lyophilized from a concentrated (1 mg/ml) sterile solution containing no additives. |
En Protein | |
Research Use Only | |
Eng | |
Recombinant Protein | |
Baculovirus | |
MDRGVLPLPITLLFVIYSFVPTTGLAERVGCDLQPVDPTRGEVT FTTSQVSEGCVAQAANAVREVHVLFLDFPGMLSHLELTLQASKQNGTETQEVF LVLVSNKNVFVKFQAPEIPLHLAYDSSLVIFQGQPRVNITVLPSLTSRKQILDWA ATKGAITSIAALDDPQSIVLQLGQDPKAPFLCLPEAHKDMGATLEWQPRAQTP VQSCRLEGVSGHKEAYILRILPGSEAGPRTVTVMMELSCTSGDAILILHGPPYVS WFIDINHSMQILTTGEYSVKIFPGSKVKGVELPDTPQGLIAEARKLNASIVTSFV ELPLVSNVSLRASSCGGVFQTTPAPVVTTPPKDTCSPVLLMSLIQPKCGNQVMT LALNKKHVQTLQCTITGLTFWDSSCQAEDTDDHLVLSSAYSSCGMKVTAHVV SNEVIISFPSGSPPLRKKVQCIDMDSLSFQLGLYLSPHFLQASNTIELGQQAFVQV SVSPLTSEVTVQLDSCHLDLGPEGDMVELIQSRTAKGSCVTLLSPSPEGDPRFSF LLRVYMVPTPTAGTLSCNLALRPSTLSQEVYKTVSMRLNIVSPDLS | |
Greater than 95.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |