missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse MIP-3 beta / CCL19 Protein
Recombinant Protein
Marke: enQuireBio™ QP5219-500ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
MIP-3 beta / CCL19 Protein | |
Research Use Only | |
Scya19 | |
Mouse | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |
500 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
GANDAEDCCL SVTQRPIPGN IVKAFRYLLN EDGCRVPAVV FTTLRGYQLC APPDQPWVDR IIRRLKKSSA KNKGNSTRRS PVS | |
Greater than 95% as determined by SDS-PAGE. |