missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse RANKL / OPGL / TNFSF11 / CD254 Protein
Recombinant Protein
Marke: enQuireBio™ QP5402-100ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
NP_035743.2 | |
RANKL / OPGL / TNFSF11 / CD254 Protein | |
Research Use Only | |
TNFSF11 | |
Recombinant Protein | |
E. coli | |
MPAMMEGSWL DVAQRGKPEA QPFAHLTINA ASIPSGSHKV TLSSWYHDRG WAKISNMTLS NGKLRVNQDG FYYLYANICF RHHETSGSVP TDYLQLMVYV VKTSIKIPSS HNLMKGGSTK NWSGNSEFHF YSINVGGFFK LRAGEEISIQ VSNPSLLDPD QDATYFGAFK VQDID | |
Greater than 95% as determined by SDS-PAGE. |
21943 | |
100 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Mouse RANKL / OPGL / TNFSF11 / CD254 induces TNF-alpha production in RAW cells (typical ED50 is 50-75 ng/ml). Activity was further confirmed by measuring Osteoclast formation (significanty different from control at Mouse RANKL / OPGL / TNFSF11 / CD254 concentrations greater than 50 ng/ml). | |
Mouse | |
Untagged | |
10 mM sodium phosphate and 20 mM sodium chloride, pH 7.5 |