Learn More
enQuireBio™ Recombinant other Sincalide Protein
Recombinant Protein
Marke: enQuireBio™ QP13506-1mg
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
Sincalide Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
The Sincalide was lyophilized from a concentrated (1 mg/ml) solution with no additives. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
Greater than 99.0% as determined by RP-HPLC. |