missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Pig Calpain-2 catalytic subunit Protein
A DNA sequence encoding the Sus scrofa (Pig) Calpain-2 catalytic subunit, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP5757-ec-200ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
Calpain-2 catalytic subunit Protein | |
Research Use Only | |
CAPN2 | |
Recombinant Protein | |
E. coli | |
YPNTFWMNPQ YLIKLEEEDE DQEDGESGCT FLVGLIQKHR RRQRKMGEDM HTIGFGIYEV PEELTGQTNI HLSKNFFLTH RARERSDTFI NLREVLNRFK LPPGEYILVP STFEPNKDGD FCIRVFSEKK ADYQVVDDEI EADLEENDAS EDDIDDGFRR LFAQLAGEDA EISAFELQTI LRRVLAKRQD IKSDGFSIET CRIMVDMLDS DGSAKLGLKE FYILWTKIQK YQKIYREIDV DRSGTMNSYE MRKALEEAGF KLPCQLHQVI VARFADDQLI IDFDNFVRCL VRLETLFRIS KQLDSENTGT IELDLISWLC FSVL | |
Greater than 90% as determined by SDS-PAGE. |
200 μg | |
Not determined. | |
Full Length | |
Pig | |
His-SUMO | |
Tris-based buffer, 50% glycerol |