missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Collagen alpha-1(I) chain Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Collagen alpha-1(I) chain, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP9077-ye-10ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
NP_445756.1 | |
Collagen alpha-1(I) chain Protein | |
Research Use Only | |
Col1a1 | |
Recombinant Protein | |
Yeast | |
QRGERGFPGL PGPSGEPGKQ GPSGASGERG PPGPMGPPGL AGPPGESGRE GSPGAEGSPG RDGAPGAKGD RGETGPAGPP GAPGAPGAPG PVGPAGKNGD RGETGPAGPA GPIGPAGARG PAGPQGPRGD KGETGEQGDR GIKGHRGFSG LQGPPGSPGS PGEQGPSGAS GPAGPRGPPG SAGSPGKDGL NGLPGPIGPP GPRGRTGDSG PAGPPGPPGP PGPPGPPSGG YDFSFLPQPP QEKSQDGGRY YRA | |
Greater than 90% as determined by SDS-PAGE. |
29393 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |