missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat FABP1 Protein
A cDNA sequence encoding the FABP1 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP11831-5ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
FABP1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMNFSGKY QVQSQENFEP FMKAMGLPED LIQKGKDIKG VSEIVHEGKK VKLTITYGSK VIHNEFTLGE ECELETMTGE KVKAVVKMEG DNKMVTTFKG IKSVTEFNGD TITNTMTLGD IVYKRVSKRI | |
Greater than 95.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Rat | |
His | |
The FABP1 solution (1 mg/1ml) contains phosphate buffered saline (pH 7.4), 20% glycerol and 1mM DTT. |