missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat IL6 / Interleukin-6 Protein
Recombinant Protein
Marke: enQuireBio™ QP5501-1mg
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
NP_036721.1 | |
IL6 / Interleukin-6 Protein | |
Research Use Only | |
IL6 | |
Recombinant Protein | |
E. coli | |
MFPTSQVRRG DFTEDTTHNR PVYTTSQVGG LITYVLREIL EMRKELCNGN SDCMNSDDAL SENNLKLPEI QRNDGCFQTG YNQEICLLKI CSGLLEFRFY LEFVKNNLQD NKKDKARVIQ SNTETLVHIF KQEIKDSYKI VLPTPTSNAL LMEKLESQKE WLRTKTIQLI LKALEEFLKV TMRSTRQT | |
Greater than 90 as determined by SDS-PAGE. |
24498 | |
1 mg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Rat IL6 / Interleukin-6 induces 7TD1 cell proliferation (typical ED50 is ? 10 pg/ml). | |
Rat | |
Untagged | |
0.1% Trifluoroacetic Acid (TFA) + 2:1 mannitol to protein |