Learn More
Thermo Scientific™ Remel™ OF Medium mit 1 % Dextrose (Hugh-Liefson)
Thermo Scientific™ Remel OF Medium dient zur Bestimmung des oxidativen und fermentativen Metabolismus von Kohlenhydraten durch gramnegative Bakterien.
274.00 €
Spezifikation
Zertifizierung/Konformität | Industrieanwendung: BAM. |
---|---|
Beschreibung | OFMedium mit 1 % Dextrose (Hugh-Liefson) |
Format | 15 mm x 103 mm, Röhrchen |
Produkttyp | Medien |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15127522
|
Thermo Scientific™
R061918 |
20 x 4 ml |
Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt. Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen |
||||||
Beschreibung
Thermo Scientific™ Remel OF Medium dient zur Bestimmung des oxidativen und fermentativen Metabolismus von Kohlenhydraten durch gramnegative Bakterien.
Typical fermentation media contain approximately 2% peptone and 1% carbohydrate. In media containing high concentrations of peptones, oxidative microorganisms produce alkaline by-products in quantities that neutralize the acid produced from carbohydrate metabolism.
OF Medium is a semisolid medium with a low peptone to carbohydrate ratio that contains the pH indicator bromthymol blue. Organisms which metabolize carbohydrate in OF medium produce acid in quantities sufficient to prevent its neutralization by alkaline peptone by-products.
- Ready to use – Convenience of prepared media
- Uses 4 mL of 1% Dextrose as carbohydrate source
- Easy to interpret – Color change from green to yellow indicates positive test for acid production.
When testing for motility, growth diffused away from stab line of inoculation indicates a positive test whereas growth confined to stab line indicates negative test.
Acid production in the open tube (without mineral oil overlay) indicates oxidative metabolism whereas acid production in both (with and without mineral oil overlay) tubes indicates fermentative metabolism
OF Medium with Dextrose uses 4 mL of 1% dextrose as a fermentable carbohydrate. The medium contains casein peptone as the substrate and bromthymol blue as the pH indicator. Dipotassium phosphate promotes fermentation and also serves as a buffer. Agar allows for detection of motility and aids in even distribution of acid produced at the surface of the medium.
Dextrose is routinely used in OF Medium; however, some organisms may be unable to metabolize dextrose but can utilize other carbohydrates. Organisms that only oxidise glucose will not ferment any other carbohydrate1. Therefore, when testing with other carbohydrates, the mineral oil overlay tube may be omitted.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Spezifikation
Industrieanwendung: BAM. | |
OFMedium mit 1 % Dextrose (Hugh-Liefson) | |
15 mm x 103 mm, Röhrchen | |
Medien |