Learn More
Restek™ Replacement Chemical Traps for Agilent™ Gas Chromatographs
Easy to install replacement chemical traps attach to the same fittings as original Agilent GC equipment
Marke: Restek™ 23032
Weitere Details : Netto-Gewicht : 0.50000kg
Beschreibung
- Built-in frits and adsorbents remove both moisture and hydrocarbons
- Trap can be regenerated to remove contaminants and restore performance
- Also available is a Replacement 32 in. Split Line with all installation hardware included
Spezifikation
Replacement Trap | |
Gas Chromatography | |
Agilent GCs |