Learn More
Restek™ Presents Hamilton PTFE Fluorocarbon Resin-Tip, Gas-Tight Syringes: Needle Sold Separately
Suitable for gases or liquids, for maximum inertness.
Marke: Restek™ 24578
Weitere Details : Netto-Gewicht : 0.10000kg
Beschreibung
- High accuracy of dispensed volumes
- Interchangeable barrels, plungers, and tips extend performance and increase cost-effectiveness
- Without slots
Spezifikation
PTFE (Tip) | |
Hamilton PTFE Tip, Gas-Tight Syringes | |
Ea. | |
No Slots |
Suitable for gases or liquids for maximum inertness | |
1001 | |
1 mL | |
Luer Lock |