Learn More
ACROBiosystems Rhesus macaque LAIR-1 / CD305 Protein, Mouse IgG2a Fc Tag
Marke: ACROBiosystems LA1-C52A3-100UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Leukocyte-associated immunoglobulin-like receptor-1 (LAIR-1) is constitutively expressed on the majority of human peripheral blood mononuclear leukocytes. LAIR-1 or CD305 is a transmembrane glycoprotein with a single immunoglobulin-like domain and a cytoplasmic tail containing two immune receptor tyrosine-based inhibitory motifs. LAIR-1 recruits SHP-1 and SHP-2 phosphatases upon activation, and cross-linking of the LAIR-1 antigen on natural killer (NK) cells results in strong inhibition of NK cell–mediated cytotoxicity. Functions as an inhibitory receptor that plays a constitutive negative regulatory role on cytolytic function of natural killer (NK) cells, B-cells and T-cells. Activation by Tyr phosphorylation results in recruitment and activation of the phosphatases PTPN6 and PTPN11. It also reduces the increase of intracellular calcium evoked by B-cell receptor ligation.Diseases associated with LAIR1 include Chronic Active Epstein-Barr Virus Infection and Palindromic Rheumatism.
This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 42.9 kDa. The protein migrates as 64 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
42.9 kDa | |
Rhesus macaque LAIR-1, Mouse IgG2a Fc Tag (LA1-C52A3) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Try 165 (Accession # ALV66454.1). | |
-20°C | |
LAIR-1 | |
Rhesus Macaque | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Immune Checkpoint Proteins |
100 μg | |
Gln 22 - Try 165 | |
LAIR1,CD305 | |
Unconjugated | |
Mouse IgG Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |