missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carl Roth™ Rotilabo™ LDPE Safety Wash Bottles
Click to view available options
Kapazität (metrisch):
500 mL
1000 mL
Chemical Label:
Distilled Water
Ethanol
Isopropanol
Beschreibung
- Bottle made of LDPE or polypropylene
- Screw cap and spray tube made of polypropylene
- Pressure relief valve in the screw cap stops discharge, dripping or leakage of liquid through the spray tube, e.g. due to temperature fluctuations
- Indelible contents label in four languages (German, English, French and Spanish)
- Hazard warning label in accordance with GHS
Spezifikation
Chemical Label | Ethanol |
Produkttyp | Safety Wash Bottle |
Kapazität (metrisch) | 500 mL |
Material | LDPE |
Anzahl pro Karton | 1Each |
Anzahl pro Packung | 1Each |
Verpackung | 1Each |
Menge | 1Each |
Nominales Fassungsvermögen | 500mL |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'