missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human RPS6KB1 (p70S6K), GST Tag Recombinant Protein
Human RPS6KB1 (p70S6K), GST Tag Recombinant Protein
Marke: Invitrogen PV3815
Weitere Details : Netto-Gewicht : 0.05250kg
Beschreibung
RPS6KB1 (p70S6K) is a serine⁄threonine kinase with 2 non-identical kinase catalytic domains which phosphorylates several residues of the S6 ribosomal protein.Spezifikation
NP_003152,T412E | |
114294, 6198 | |
10 μg | |
RSK (RPS6K) Kinase Family | |
Human | |
GST Tag | |
Biochemical binding/kinetic, Biochemical phospho-specific activity (no antibody) | |
Dry Ice | |
RPS6KB1 | |
Baculovirus | |
Recombinant, Catalytic | |
Serine/Threonine Kinases |
Kinase Assay | |
75.1 kDa | |
RPS6KB1 (p70S6K) | |
aa 1-421 | |
Recombinant | |
Storage Buffer: 50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Ser⁄Thr Kinases: AGC Group | |
LanthaScreen™ Antibody, LanthaScreen™ Substrate, Z'-LYTE™, Omnia™ Bulk Substrate | |
ribosomal protein S6 kinase, 70kDa, polypeptide 1 | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction |