missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S0859, MedChemExpress
Click to view available options
Menge:
5 mg
10 mg
50 mg
missing translation for 'unitSize'
10 Milligramm
50 Milligramm
Description
S0859 is a selective, high-affinity generic Na+/HCO3- transporter (NBC) inhibitor. S0859 reversibly inhibits NBC-mediated intracellular pH (pHi) recovery (Ki=1.7 μM, full inhibition at approximately 30 μM).
Specifications
Specifications
| Chemischer Name oder Material | S0859 |
| CAS | 1019331-10-2 |
| Farbe | White |
| Hinweise zur Reinheitsqualität | Research |
| Summenformel | C29H24ClN3O3S |
| Menge | 50 mg |
| Löslichkeitsinformationen | DMSO : ≥ 100 mg/mL (188.67 mM) |
| SMILES | O=C(N(CC1=CC=C(C2=CC=CC=C2S(=O)(NC#N)=O)C=C1)CC3=CC=C(C)C=C3)C4=CC=CC=C4Cl |
| Molekulargewicht (g/mol) | 530.04 |
| Formelmasse | 530.04 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction