missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SERPINA10 Mouse anti-Human, Porcine, Rat, Clone: 5B3C9, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 60113-1-IG-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
The protein encoded by this gene belongs to the serpin family. It is predomitly expressed in the liver and secreted in plasma. It inhibits the activity of coagulation factors Xa and XIa in the presence of protein Z, calcium and phospholipid. Mutations in this gene are associated with venous thrombosis. Alternatively spliced transcript variants have been found for this gene.Spezifikation
SERPINA10 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q62975, Q9UK55 | |
SERPINA10 | |
SERPINA10 Fusion Protein Ag2420 | |
20 μL | |
Primary | |
Porcine, Human, Rat | |
Antibody | |
IgG2a |
Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
5B3C9 | |
Unconjugated | |
SERPINA10 | |
PZI, Serpin A10, SERPINA10, ZPI | |
Mouse | |
Protein A | |
RUO | |
171154, 51156 | |
-20°C | |
Liquid |