missing translation for 'onlineSavingsMsg'
Learn More

TAK-593, MedChemExpress

Product Code. 16499147
Encompass_Preferred
Change view
Click to view available options
Menge:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge
16499147 5 mg
16492667 50 mg
16486578 10 mg
16456786 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16499147 Supplier Medchem Express Supplier No. HY15506
 Eingeschränkter Artikel
Verfügbar ab: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress TAK-593 is a potent VEGFR and PDGFR family inhibitor with IC50s of 3.2, 0.95, 1.1, 4.3 and 13 nM for VEGFR1, VEGFR2, VEGFR3, PDFGRα and PDFGRβ, respectively.

TAK-593 is a potent VEGFR and PDGFR family inhibitor with IC50s of 3.2, 0.95, 1.1, 4.3 and 13 nM for VEGFR1, VEGFR2, VEGFR3, PDFGRα and PDFGRβ, respectively.

Specifications

Chemischer Name oder Material TAK-593
CAS 1005780-62-0
Farbe Light Yellow
Hinweise zur Reinheitsqualität Research
Summenformel C23H23N7O3
Menge 5 mg
Löslichkeitsinformationen DMSO : ≥ 48.5 mg/mL (108.87 mM)
SMILES O=C(C1=CC(C)=NN1C)NC2=CC(OC3=NN4C(C=C3)=NC(NC(C5CC5)=O)=C4)=CC=C2C
Molekulargewicht (g/mol) 445.47
Formelmasse 445.47
Reinheit (%) 98.0%
Güte Research
Physikalische Form Solid
Zur Verwendung mit (Anwendung) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.