missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIM-3 (CD366) Mouse anti-Human, Alexa Fluor 647, Clone: 7D3, BD Pharmingen™
Mouse Monoclonal Antibody
Marke: BD Biosciences 565558
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
TIM-3 (CD366) Monoclonal antibody specifically detects TIM-3 (CD366) in Human samples. It is validated for Flow cytometry.
TIM-3 (CD366) Monoclonal antibody specifically detects TIM-3 (CD366) in Human samples. It is validated for Flow cytometry.Spezifikation
TIM-3 (CD366) | |
Monoclonal | |
Alexa Fluor 647 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Mouse | |
100 Tests | |
Cell Biology | |
Human | |
Purified |
Flow Cytometry | |
7D3 | |
Vol. Per test: 5μL | |
CD366; HAVCR2; TIM3; T cell immunoglobulin mucin-3; TIMD-3; KIM-3 | |
Human TIM-3 | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to Alexa Fluor™ 647 under optimum conditions and unreacted Alexa Fluor™ 647 was removed. | |
IgG1 κ |
For Research Use Only.