missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIP30 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 10251-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
CC3 (HTATIP2) is a member of the short-chain dehydrogenases/reductases (SDR) family. It is a novel serine/threonine kinase that phosphorylates the C-terminal domain (CTD) of the largest RNA polymerase II subunit and induces the expression of apoptosis related genes Bad and Siva, as well as metastasis suppressor NM23-H2. It also interacts with an estrogen receptor alpha-interacting coactivator CIA and regulates ERalpha-mediated c-myc transcription.Spezifikation
TIP30 | |
Polyclonal | |
Unconjugated | |
HTATIP2 | |
CC3, HTATIP2, Oxidoreductase HTATIP2, SDR44U1, TIP30 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10553, 292935, 53415 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9BUP3, Q9Z2G9 | |
Htatip2 | |
TIP30 Fusion Protein Ag0353 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |