Learn More
TMEM119 Mouse anti-Human, Unconjugated, Clone: 1E7A2, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 66948-1-IG-150UL
Additional Details : Netto-Gewicht : 0.00010kg
Beschreibung
The TMEM119 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TMEM119. This antibody recognizes human, pig, mouse, rat antigen. The TMEM119 antibody has been validated for the following applications: WB, ELISA analysis.
Spezifikation
TMEM119 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Mouse, Rat, Pig |
Western Blot, Immunohistochemistry, Immunofluorescence | |
1E7A2 | |
Unconjugated | |
150 μL | |
338773 | |
IgG3 |