missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNNI1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 16102-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Troponin proteins associate with tropomyosin and regulate the calcium sensitivity of the myofibril contractile apparatus of striated muscles. Troponin I (TnI), along with troponin T (TnT) and troponin C (TnC), is one of 3 subunits that form the troponin complex of the thin filaments of striated muscle. TnI is the inhibitory subunit; blocking actin-myosin interactions and thereby mediating striated muscle relaxation. The TnI subfamily contains three genes: TnI-skeletal-fast-twitch, TnI-skeletal-slow-twitch, and TnI-cardiac. The TnI-fast and TnI-slow genes are expressed in fast-twitch and slow-twitch skeletal muscle fibers, respectively, while the TnI-cardiac gene is expressed exclusively in cardiac muscle tissue. This gene encodes the Troponin-I-skeletal-slow-twitch protein. This gene is expressed in cardiac and skeletal muscle during early development but is restricted to slow-twitch skeletal muscle fibers in adults. The encoded protein prevents muscle contraction by inhibiting calcium-mediated conformational changes in actin-myosin complexes.Spezifikation
TNNI1 | |
Polyclonal | |
Unconjugated | |
TNNI1 | |
SSTNI, TNN1, TNNI1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
21952, 29388, 7135 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.14 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P13413, P19237, Q9WUZ5 | |
Tnni1 | |
TNNI1 Fusion Protein Ag9001 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |