missing translation for 'onlineSavingsMsg'
Learn More

trans-4-(Benzyloxycarbonylamino)cyclohexancarboxaldehyd, 97 %, Thermo Scientific Chemicals

Catalog Number p-7041653
Click to view available options
Menge:
250 mg
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
412357-50-7
C15H19NO3
261.32
MFCD03844593,MFCD06657671
SOIRQISQWQDGSN-UHFFFAOYSA-N
21882542
benzyl N-(4-formylcyclohexyl)carbamate
O=CC1CCC(CC1)NC(=O)OCC1=CC=CC=C1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
412357-50-7
C15H19NO3
261.32
MFCD03844593,MFCD06657671
SOIRQISQWQDGSN-UHFFFAOYSA-N
21882542
benzyl N-(4-formylcyclohexyl)carbamate
O=CC1CCC(CC1)NC(=O)OCC1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material trans-4-(Benzyloxycarbonylamino)cyclohexanecarboxaldehyde
Synonym benzyl 4-formylcyclohexylcarbamate, benzyl cis-4-formylcyclohexyl carbamate, benzyl cis-4-formylcyclohexylcarbamate, benzyl 4-formylcyclohexyl carbamate, trans-4-benzyloxycarbonylamino-cyclohexanecarbaldehyde, benzyl 1r,4r-4-formylcyclohexylcarbamate, trans-4-benzyloxycarbonylamino cyclohexanecarbaldehyde, benzyl n-4-formylcyclohexyl carbamate, benzyl4-formylcyclohexylcarbamate, soirqisqwqdgsn-mqmhxkeqsa-n
Formelmasse 261.31
Reinheit (%) 97%
Menge 250 mg

RUO – Nur für Forschungszwecke

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'