missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trelagliptin succinate, MedChemExpress
Beschreibung
Trelagliptin (SYR-472) succinate is a potent, orally active and highly selective DPP-4 inhibitor with an IC50 of 4 nM. Trelagliptin succinate improves glycemic control in vivo and can be used for the study of type 2 diabetes mellitus (T2DM).
Spezifikation
Spezifikation
| Chemischer Name oder Material | Trelagliptin succinate |
| CAS | 1029877-94-8 |
| Farbe | White |
| Hinweise zur Reinheitsqualität | Research |
| Summenformel | C22H26FN5O6 |
| Menge | 100 mg |
| Synonym | SYR-472 succinate |
| Löslichkeitsinformationen | H2O : 100 mg/mL (210.32 mM; Need ultrasonic) ∣DMSO : ≥ 50 mg/mL (105.16 mM) |
| SMILES | O=C(C=C(N1C[C@H](N)CCC1)N2CC3=C(C#N)C=CC(F)=C3)N(C)C2=O.O=C(O)CCC(O)=O |
| Molekulargewicht (g/mol) | 475.47 |
| Mehr anzeigen |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?