missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TUFM Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 26730-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
This gene encodes a protein which participates in protein translation in mitochondria. Mutations in this gene have been associated with combined oxidative phosphorylation deficiency resulting in lactic acidosis and fatal encephalopathy. A pseudogene has been identified on chromosome 17.Spezifikation
TUFM | |
Polyclonal | |
Unconjugated | |
TUFM | |
COXPD4, EF Tu, EF TuMT, EFTU, P43, TUFM | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
233870, 293481, 7284 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P49411, P85834, Q8BFR5 | |
Tufm | |
TUFM Fusion Protein Ag24981 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |