missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tyrosinase Antibody (T311 + OCA1/812), Novus Biologicals™
Mouse Monoclonal Antibody
265.00 € - 474.00 €
Spezifikation
| Antigen | Tyrosinase |
|---|---|
| Klon | T311 + OCA1/812 |
| Konzentration | 0.2mg/mL |
| Verdünnung | Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, Immunofluorescence 1 - 2 ug/ml |
| Anwendungen | Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
18199308
|
Novus Biologicals
NBP2-44686-0.02MG |
0.02 mg |
265.00 €
0.02 Milligramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
18146269
|
Novus Biologicals
NBP2-44686-0.1MG |
0.1 mg |
474.00 €
0.10 Milligramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
Ensure accurate, reproducible results in Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence
Tyrosinase Monoclonal specifically detects Tyrosinase in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence.Spezifikation
| Tyrosinase | |
| 0.2mg/mL | |
| Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Lipid and Metabolism | |
| 10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
| 7299 | |
| Recombinant tyrosinase protein (T311); Recombinant human TYR protein (OCA1/812) | |
| Primary | |
| Store at 4C. | |
| 75 kDa |
| T311 + OCA1/812 | |
| Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, Immunofluorescence 1 - 2 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CMM8, EC 1.14.18.1, LB24-AB, Monophenol monooxygenase, OCA1A, OCAIA, SHEP3, SK29-AB, Tumor rejection antigen AB, tyrosinase, tyrosinase (oculocutaneous albinism IA) | |
| TYR | |
| IgG | |
| Protein A or G purified | |
| Recognizes a cluster of proteins between 70-80kDa, identified as tyrosinase. Occasionally a minor band at 55kDa is also detected. This MAb shows no cross-reaction with MAGE-1 and tyrosinase-related protein 1, TRP-1/gp75. Tyrosinase is a copper-containing metalloglycoprotein that catalyzes several steps in the melanin pigment biosynthetic pathway; the hydroxylation of tyrosine to L-3,4-dihydroxy-phenylalanine (dopa), and the subsequent oxidation of dopa to dopaquinone. Mutations of the tyrosinase gene occur in various forms of albinism. Tyrosinase is one of the targets for cytotoxic T-cell recognition in melanoma patients. Staining of melanomas with this MAb shows tyrosinase in melanotic as well as amelanotic variants. This MAb is a useful marker for melanocytes and melanomas. |
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts