missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UCKL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 17005-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
The protein encoded by this gene is a uridine kinase. Uridine kinases catalyze the phosphorylation of uridine to uridine monophosphate. This protein has been shown to bind to Epstein-Barr nuclear antigen 3 as well as natural killer lytic-associated molecule. Ubiquitination of this protein is enhanced by the presence of natural killer lytic-associated molecule. In addition, protein levels decrease in the presence of natural killer lytic-associated molecule, suggesting that association with natural killer lytic-associated molecule results in ubiquitination and subsequent degradation of this protein. Alternative splicing results in multiple transcript variants.Spezifikation
UCKL1 | |
Polyclonal | |
Unconjugated | |
UCKL1 | |
UCK1 LIKE, UCK1L, UCKL1, Uridine cytidine kinase like 1, URKL1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
499956, 54963, 68556 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q91YL3, Q9NWZ5 | |
UCKL1 | |
UCKL1 Fusion Protein Ag10701 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |