missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WRN Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 25269-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
This gene encodes a member of the RecQ subfamily and the DEAH (Asp-Glu-Ala-His) subfamily of DNA and RNA helicases. DNA helicases are involved in many aspects of DNA metabolism, including transcription, replication, recombination, and repair. This protein contains a nuclear localization signal in the C-terminus and shows a predomit nucleolar localization. It possesses an intrinsic 3′ to 5′ DNA helicase activity, and is also a 3′ to 5′ exonuclease. Based on interactions between this protein and Ku70/80 heterodimer in DNA end processing, this protein may be involved in the repair of double strand DNA breaks. Defects in this gene are the cause of Werner syndrome, an autosomal recessive disorder characterized by premature aging.Spezifikation
WRN | |
Polyclonal | |
Unconjugated | |
WRN | |
DNA helicase, RecQ like type 3, Exonuclease WRN, RecQ protein like 2, RECQ3, RECQL2, RECQL3, WRN | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
7486 | |
-20°C | |
Liquid |
Western Blot | |
0.56 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q14191 | |
WRN | |
Peptide | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |