missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YE 120, Tocris Bioscience™
Click to view available options
Menge:
10 mg
50 mg
Beschreibung
GPR35 agonist (EC50 = 32 nM in DMR assays). Also displays partial agonist activity in a β-arrestin translocation assay (EC50 = 10.2 μM).
Spezifikation
Chemischer Name oder Material | YE 120 |
Menge | 10 mg |
Formelmasse | 330.17 |
Reinheit (%) | >98% |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'