CAS RN 2443816-45-1
CAS RN 2443816-45-1
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
1
von
1
Ergebnisse
1
ULK-101, MedChemExpress
MedChemExpress ULK-101 is a potent and selective ULK1 inhibitor, with IC50 values of 1.6 nM and 30 nM for ULK1 and ULK2, respectively. ULK-101 suppresses autophagy and sensitizes cancer cells to nutrient stress.