CAS RN 7803-88-5
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
2
von
2
Ergebnisse
1
6-O-Methylguanosin, TRC
CAS: 7803-88-5 Summenformel: C11 H15 N5 O5 Molekulargewicht (g/mol): 297.27 IUPAC-Name: (2R,3R,4S,5R)-2-(2-Amino-6-Methoxypurin-9-yl)-5-(Hydroxymethyl)Oxol-3,4-Diol SMILES: COc1nc(N)nc2c1ncn2[C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O
2
6-O-Methyl Guanosine, MedChemExpress
MedChemExpress 6-O-Methyl Guanosine is a modified nucleoside. 6-O-Methyl Guanosine (6-methylguanosine) inhibit colony-forming ability in a malignant xeroderma pigmentosum cell line.