CAS RN 90098-04-7
CAS RN 90098-04-7
IUPAC Name:
2-[(4-chlorophenyl)formamido]-3-(2-oxo-1,2-dihydroquinolin-4-yl)propanoic acid
Synonyme:
rebamipide
Molekulargewicht (g/mol):
370.79
Summenformel:
C19H15ClN2O4
InChi Key:
ALLWOAVDORUJLA-UHFFFAOYNA-N
SMILES:
OC(=O)C(CC1=CC(=O)NC2=CC=CC=C12)NC(=O)C1=CC=C(Cl)C=C1
1
–
2
von
2
Ergebnisse
1
Rebamipide, TRC
CAS: 90098-04-7 Summenformel: C19H15ClN2O4 Molekulargewicht (g/mol): 370.79 Synonym: 4-Quinolinepropanoic acid, α-[(4-chlorobenzoyl)amino]-1,2-dihydro-2-oxo-,α-[(4-Chlorobenzoyl)amino]-1,2-dihydro-2-oxo-4-quinolinepropanoic acid,Mucosta,OPC 12759,Proamipide,Rebamipide,Rebator IUPAC-Name: 2-[(4-chlorobenzoyl)amino]-3-(2-oxo-1H-quinolin-4-yl)propanoic acid SMILES: OC(=O)C(CC1=CC(=O)Nc2ccccc12)NC(=O)c3ccc(Cl)cc3
2
Rebamipide, MedChemExpress
MedChemExpress Rebamipide (OPC12759) is a mucoprotective agent. Rebamipide induces COX-2 expression, increases PGE2 levels, and enhances gastric mucosal defense in a COX-2-dependent manner.