CAS RN 1000787-75-6
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
1
von
1
Ergebnisse
1
Tegobuvir, MedChemExpress
MedChemExpress Tegobuvir is a specific, covalent inhibitor of the HCV NS5B polymerase.