CAS RN 100299-08-9
IUPAC Name:
potassium 5-{9-methyl-4-oxo-4H-pyrido[1,2-a]pyrimidin-3-yl}-1H-1,2,3,4-tetrazol-1-ide
Synonyme:
pemirolast
Molekulargewicht (g/mol):
266.31
Summenformel:
C10H7KN6O
InChi Key:
NMMVKSMGBDRONO-UHFFFAOYSA-N
SMILES:
[K+].CC1=CC=CN2C(=O)C(=CN=C12)C1=NN=N[N-]1
1
–
2
von
2
Ergebnisse
2
Pemirolast potassium, MedChemExpress
MedChemExpress Pemirolast potassium (TWT-8152) is a histamine H1 antagonist and mast cell stabilizer that acts as an antiallergic agent.