CAS RN 1010085-13-8
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
1
von
1
Ergebnisse
1
MK-5108, MedChemExpress
MedChemExpress MK-5108 is a highly potent and specific inhibitor of Aurora A kinase with an IC50 value of 0.064 nM.