CAS RN 1012-05-1
CAS RN 1012-05-1
IUPAC Name:
2-amino-4-phenylbutanoic acid
Synonyme:
homophenylalanine
Molekulargewicht (g/mol):
179.22
Summenformel:
C10H13NO2
InChi Key:
JTTHKOPSMAVJFE-UHFFFAOYNA-N
SMILES:
NC(CCC1=CC=CC=C1)C(O)=O
1
–
1
von
1
Ergebnisse
1
(2RS)-2-Amino-4-phenylbutanoic Acid, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.