CAS RN 101831-37-2
CAS RN 101831-37-2
IUPAC Name:
2-(4-chlorophenyl)-2-[2,6-dichloro-4-(3,5-dioxo-2,3,4,5-tetrahydro-1,2,4-triazin-2-yl)phenyl]acetonitrile
Synonyme:
diclazuril
Molekulargewicht (g/mol):
407.64
Summenformel:
C17H9Cl3N4O2
InChi Key:
ZSZFUDFOPOMEET-UHFFFAOYNA-N
SMILES:
ClC1=CC=C(C=C1)C(C#N)C1=C(Cl)C=C(C=C1Cl)N1N=CC(=O)NC1=O
1
–
1
von
1
Ergebnisse
1
Diclazuril, MedChemExpress
MedChemExpress Diclazuril (R-64433), a benzeneacetonitrile derivative, is a potent and orally active anticoccidial agent. Diclazuril can be used for the research of certain infectious and parasitic diseases, including coccidiosis, acute toxoplasmosis, equine protozoal pyoencephalitis (EPM) et.al.
