CAS RN 1021497-97-1
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
1
von
1
Ergebnisse
1
KR-33493, MedChemExpress
MedChemExpress KR-33493 is a potent inhibitor of Fas-mediated cell death (FAF1).