CAS RN 1021950-26-4
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
1
von
1
Ergebnisse
1
Tyrosine kinase inhibitor, MedChemExpress
MedChemExpress Tyrosine kinase inhibitor is a potent tyrosine kinase inhibitor.