CAS RN 1032754-81-6
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
1
von
1
Ergebnisse
1
GNE-477, MedChemExpress
MedChemExpress GNE-477 is a potent and efficacious dual PI3K (IC50=4 nM)/mTOR(Ki=21 nM) inhibitor.