CAS RN 1268244-88-7
CAS RN 1268244-88-7
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
1
von
1
Ergebnisse
1
(-)-PX20606 trans isomer, MedChemExpress
MedChemExpress (-)-PX20606 trans isomer is a FXR agonist with EC50s of 18 and 29 nM for FXR in FRET and M1H assay, respectively.