CAS RN 1584713-87-0
CAS RN 1584713-87-0
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
1
von
1
Ergebnisse
1
BMS-983970, MedChemExpress
MedChemExpress BMS-983970 is an oral pan-Notch inhibitor for the treatment of multiplecancers.