CAS RN 204697-65-4
CAS RN 204697-65-4
IUPAC Name:
N-{6-amino-1-oxo-1-[4-(pyridin-4-yl)piperazin-1-yl]hexan-2-yl}-3-(3,5-dibromo-4-hydroxyphenyl)-2-{[4-(2-oxo-1,2,3,4-tetrahydroquinazolin-3-yl)piperidine-1-carbonyl]amino}propanamide
Synonyme:
olcegepant
Molekulargewicht (g/mol):
869.66
Summenformel:
C38H47Br2N9O5
InChi Key:
ITIXDWVDFFXNEG-UHFFFAOYNA-N
SMILES:
NCCCCC(NC(=O)C(CC1=CC(Br)=C(O)C(Br)=C1)NC(=O)N1CCC(CC1)N1CC2=CC=CC=C2NC1=O)C(=O)N1CCN(CC1)C1=CC=NC=C1
1
–
2
von
2
Ergebnisse
1
Tocris Bioscience™ BIBN 4096
Potent and selective CGRP antagonist
2
Olcegepant, MedChemExpress
MedChemExpress Olcegepant (BIBN-4096) is a potent and selective non-peptide antagonist of the calcitonin gene-related peptide 1 (CGRP1) receptor with IC50 of 0.03 nM and Ki of 14.4 pM for human CGRP.