CAS RN 2070009-61-7
CAS RN 2070009-61-7
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
3
von
3
Ergebnisse
1
MedChemExpress Z-Gly-Gly-Arg-AMC acetate
MedChemExpress Z-Gly-Gly-Arg-AMC acetate is a thrombin-specific fluorogenic substrate for testing of thrombin generation in PRP and platelet-poor plasma (PPP).
2
MedChemExpress Z-Gly-Gly-Arg-AMC acetate
MedChemExpress Z-Gly-Gly-Arg-AMC acetate is a thrombin-specific fluorogenic substrate for testing of thrombin generation in PRP and platelet-poor plasma (PPP).
3
MedChemExpress Z-Gly-Gly-Arg-AMC acetate
MedChemExpress Z-Gly-Gly-Arg-AMC acetate is a thrombin-specific fluorogenic substrate for testing of thrombin generation in PRP and platelet-poor plasma (PPP).